The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024731.3 | NP_079007.2 | 4 - 5 | 1453 | 90 |
XM_005256149.2 | XP_005256206.1 | 4 - 5 | 1667 | 90 | |
XM_017023674.1 | XP_016879163.1 | 4 - 5 | 1415 | 90 | |
XM_017023675.1 | XP_016879164.1 | 4 - 5 | 1435 | 90 | |
GenBank mRNA | AK022605.1 | - | 3 - 4 | 1306 | 90 |
AK074056.1 | - | 1 - 2 | 382 | 90 | |
AK122745.1 | - | 2 - 3 | 1888 | 90 | |
BC033821.1 | - | 4 - 5 | 1439 | 90 |