Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDB1
Entrez Gene ID
23187
Gene Name
pleckstrin homology like domain family B member 1
Gene Aliases
LL5A
Chromosome Location
Chr.11: 118606296 - 118658038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144758.2 NP_001138230.1 20 - 21 4004 59
NM_001144759.2 NP_001138231.1 19 - 20 3863 59
NM_015157.3 NP_055972.1 21 - 22 4227 59
XM_005271468.3 XP_005271525.1 21 - 22 4818 59
XM_005271469.3 XP_005271526.1 21 - 22 4818 59
XM_005271470.3 XP_005271527.1 20 - 21 4773 59
XM_005271471.3 XP_005271528.1 20 - 21 4773 59
XM_005271472.3 XP_005271529.1 20 - 21 4677 59
XM_005271473.3 XP_005271530.1 19 - 20 4632 59
XM_005271474.4 XP_005271531.1 19 - 20 4632 59
XM_005277680.3 XP_005277737.1 24 - 25 5259 59
XM_006718796.3 XP_006718859.1 24 - 25 4448 59
XM_006718797.3 XP_006718860.1 23 - 24 5118 59
XM_006718798.3 XP_006718861.1 22 - 23 4947 59
XM_006718799.3 XP_006718862.1 21 - 22 4902 59
XM_006718800.3 XP_006718863.1 21 - 22 4902 59
XM_006718801.3 XP_006718864.1 21 - 22 4806 59
XM_006718802.3 XP_006718865.1 20 - 21 4761 59
XM_006718803.3 XP_006718866.1 20 - 21 4761 59
XM_011542703.2 XP_011541005.1 24 - 25 5262 59
XM_011542704.2 XP_011541006.1 24 - 25 5262 59
XM_011542705.2 XP_011541007.1 23 - 24 5217 59
XM_011542706.2 XP_011541008.1 23 - 24 5217 59
XM_011542707.2 XP_011541009.1 23 - 24 5133 59
XM_011542708.2 XP_011541010.1 24 - 25 4451 59
XM_011542709.2 XP_011541011.1 24 - 25 4761 59
XM_011542710.2 XP_011541012.1 23 - 24 5091 59
XM_011542711.2 XP_011541013.1 21 - 22 4168 59
XM_011542713.2 XP_011541015.1 21 - 22 4162 59
XM_011542715.2 XP_011541017.1 14 - 15 3139 59
XM_017017407.1 XP_016872896.1 23 - 24 5130 59
XM_017017408.1 XP_016872897.1 22 - 23 5073 59
XM_017017409.1 XP_016872898.1 22 - 23 5046 59
XM_017017410.1 XP_016872899.1 22 - 23 4989 59
GenBank mRNA AB014538.2 - 20 - 21 3981 59
AB094090.1 - 21 - 22 4227 59
AB177859.1 - 16 - 17 3153 59
AK027510.1 - 11 - 12 1492 59
AK074070.2 - 19 - 20 3812 59
AK074747.1 - 5 - 6 747 59
AK074918.1 - 13 - 14 1699 59
AK125783.1 - 22 - 23 3231 59
AL832373.1 - 18 - 19 3355 59
BC013031.2 - 13 - 14 1849 59
BC014510.1 - 6 - 7 510 59
BC098586.1 - 19 - 20 4357 59
CR749783.1 - 20 - 21 4520 59

View Full Product Details