The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025248.1 | NP_001020419.1 | 3 - 4 | 603 | 87 |
NM_001025249.1 | NP_001020420.1 | 3 - 4 | 287 | 87 | |
NM_001948.3 | NP_001939.1 | 2 - 3 | 331 | 87 | |
XM_011521315.2 | XP_011519617.1 | 3 - 4 | 323 | 87 | |
GenBank mRNA | AB049113.2 | - | 2 - 3 | 266 | 87 |
AK000629.1 | - | 3 - 4 | 283 | 87 | |
AK291515.1 | - | 2 - 3 | 87 | ||
AK312122.1 | - | 2 - 3 | 299 | 87 | |
BC033645.1 | - | 2 - 3 | 359 | 87 | |
BC070339.1 | - | 3 - 4 | 587 | 87 | |
BC110377.1 | - | 3 - 4 | 582 | 87 | |
BG505144.1 | - | 3 - 4 | 303 | 87 | |
BG532018.1 | - | - | 333 | 87 | |
CR541720.1 | - | 2 - 3 | 247 | 87 | |
CR541781.1 | - | 2 - 3 | 247 | 87 | |
L11877.1 | - | 2 - 3 | 267 | 87 | |
M89913.1 | - | 2 - 3 | 265 | 87 | |
U31930.1 | - | 2 - 3 | 276 | 87 | |
U62891.1 | - | 2 - 3 | 278 | 87 | |
U90223.1 | - | 3 - 4 | 573 | 87 |