Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr.1: 234391313 - 234479174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005646.3 NP_005637.3 10 - 11 1863 146
XM_011544263.2 XP_011542565.1 10 - 11 1930 146
XM_017002194.1 XP_016857683.1 10 - 11 1933 146
XM_017002195.1 XP_016857684.1 7 - 8 736 146
XM_017002197.1 XP_016857686.1 7 - 8 736 146
GenBank mRNA U38847.1 - 10 - 11 1863 146

View Full Product Details