Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr.1: 234391313 - 234479174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005646.3 NP_005637.3 22 - 23 3708 155
XM_005273234.3 XP_005273291.1 22 - 23 3705 155
XM_011544263.2 XP_011542565.1 21 - 22 3703 155
XM_017002195.1 XP_016857684.1 19 - 20 2581 155
XM_017002196.1 XP_016857685.1 10 - 11 1553 155
XM_017002197.1 XP_016857686.1 17 - 18 2321 155
GenBank mRNA U38847.1 - 22 - 23 3708 155

View Full Product Details