Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARBP1
Entrez Gene ID
6894
Gene Name
TAR (HIV-1) RNA binding protein 1
Gene Aliases
TRM3, TRP-185, TRP185
Chromosome Location
Chr.1: 234391313 - 234479174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005646.3 NP_005637.3 24 - 25 3988 93
XM_005273234.3 XP_005273291.1 24 - 25 3985 93
XM_011544263.2 XP_011542565.1 23 - 24 3983 93
XM_017002195.1 XP_016857684.1 21 - 22 2861 93
XM_017002196.1 XP_016857685.1 12 - 13 1833 93
XM_017002197.1 XP_016857686.1 19 - 20 2601 93
GenBank mRNA AK055044.1 - 1 - 2 2056 93
BC016023.1 - 1 - 2 481 93
U38847.1 - 24 - 25 3988 93

View Full Product Details