The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001980.3 | NP_001971.2 | 2 - 3 | 287 | 148 |
NM_194356.2 | NP_919337.1 | 2 - 3 | 287 | 148 | |
XM_005253557.1 | XP_005253614.1 | 2 - 3 | 287 | 148 | |
XM_017018978.1 | XP_016874467.1 | 2 - 3 | 284 | 148 | |
XM_017018979.1 | XP_016874468.1 | 2 - 3 | 284 | 148 | |
XM_017018980.1 | XP_016874469.1 | 2 - 3 | 280 | 148 | |
XM_017018981.1 | XP_016874470.1 | 2 - 3 | 289 | 148 | |
XM_017018982.1 | XP_016874471.1 | 2 - 3 | 284 | 148 | |
XM_017018983.1 | XP_016874472.1 | 2 - 3 | 228 | 148 | |
XM_017018985.1 | XP_016874474.1 | 2 - 3 | 246 | 148 | |
XM_017018986.1 | XP_016874475.1 | 2 - 3 | 303 | 148 | |
XM_017018987.1 | XP_016874476.1 | 2 - 3 | 305 | 148 | |
XM_017018988.1 | XP_016874477.1 | 2 - 3 | 287 | 148 | |
XM_017018989.1 | XP_016874478.1 | 2 - 3 | 351 | 148 | |
GenBank mRNA | BC047496.1 | - | 2 - 3 | 279 | 148 |
D14582.1 | - | 2 - 3 | 207 | 148 | |
DB474363.1 | - | 2 - 3 | 287 | 148 |