The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035247.2 | NP_001030324.2 | 9 - 10 | 68 | |
NM_005659.6 | NP_005650.2 | 9 - 10 | 68 | ||
XM_011530361.2 | XP_011528663.1 | 9 - 10 | 68 | ||
GenBank mRNA | AF141201.1 | - | 8 - 9 | 68 | |
AJ239058.1 | - | 8 - 9 | 68 | ||
AK128120.1 | - | 4 - 5 | 68 | ||
AK225877.1 | - | 9 - 10 | 68 | ||
AK304734.1 | - | 9 - 10 | 68 | ||
AK315998.1 | - | 9 - 10 | 68 | ||
AY101594.1 | - | 9 - 10 | 68 | ||
BC001049.2 | - | 9 - 10 | 68 | ||
BC005087.1 | - | 9 - 10 | 68 | ||
BX648483.1 | - | 9 - 10 | 68 | ||
CR456607.1 | - | 9 - 10 | 68 | ||
U64444.1 | - | 9 - 10 | 865 | 68 |