The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199917.1 | NP_001186846.1 | 5 - 6 | 712 | 130 |
NM_001199918.1 | NP_001186847.1 | 3 - 4 | 370 | 130 | |
NM_001199919.1 | NP_001186848.1 | 4 - 5 | 576 | 130 | |
NM_015599.2 | NP_056414.1 | 4 - 5 | 576 | 130 | |
XM_017010935.1 | XP_016866424.1 | 3 - 4 | 343 | 130 | |
XM_017010936.1 | XP_016866425.1 | 4 - 5 | 575 | 130 | |
XM_017010937.1 | XP_016866426.1 | 3 - 4 | 343 | 130 | |
XM_017010938.1 | XP_016866427.1 | 4 - 5 | 564 | 130 | |
GenBank mRNA | AB032081.1 | - | 4 - 5 | 488 | 130 |
AF102265.1 | - | 4 - 5 | 492 | 130 | |
AF180371.1 | - | 4 - 5 | 499 | 130 | |
AK023432.1 | - | 4 - 5 | 521 | 130 | |
AK225363.1 | - | 4 - 5 | 514 | 130 | |
AK301867.1 | - | 5 - 6 | 690 | 130 | |
AK314512.1 | - | 4 - 5 | 509 | 130 | |
BC001258.1 | - | 4 - 5 | 535 | 130 |