The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004181.4 | NP_004172.2 | 3 - 4 | 272 | 72 |
GenBank mRNA | AK054579.1 | - | 2 - 3 | 707 | 72 |
AK315368.1 | - | 3 - 4 | 221 | 72 | |
BC000332.2 | - | 3 - 4 | 241 | 72 | |
BC005117.1 | - | 3 - 4 | 200 | 72 | |
BC006305.2 | - | 3 - 4 | 212 | 72 | |
BC018838.2 | - | 3 - 4 | 241 | 72 | |
BP202166.1 | - | 3 - 4 | 272 | 72 | |
FJ224355.1 | - | 3 - 4 | 241 | 72 | |
GQ472201.1 | - | 3 - 4 | 212 | 72 | |
X04741.1 | - | 3 - 4 | 174 | 72 |