The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 24 - 25 | 2285 | 143 |
NM_001282542.1 | NP_001269471.1 | 24 - 25 | 2285 | 143 | |
NM_003176.3 | NP_003167.2 | 24 - 25 | 2282 | 143 | |
NR_104211.1 | 24 - 25 | 2282 | 143 | ||
XM_005271155.3 | XP_005271212.1 | 23 - 24 | 2606 | 143 | |
XM_005271156.1 | XP_005271213.1 | 23 - 24 | 2080 | 143 | |
XM_006710859.1 | XP_006710922.1 | 24 - 25 | 2193 | 143 | |
XM_011542037.2 | XP_011540339.1 | 24 - 25 | 2249 | 143 | |
XM_011542038.2 | XP_011540340.1 | 17 - 18 | 1476 | 143 | |
XM_017002184.1 | XP_016857673.1 | 24 - 25 | 2085 | 143 | |
XM_017002185.1 | XP_016857674.1 | 18 - 19 | 1511 | 143 | |
GenBank mRNA | AF273043.1 | - | 24 - 25 | 2220 | 143 |
AK315268.1 | - | 24 - 25 | 2244 | 143 | |
BC126266.1 | - | 24 - 25 | 2159 | 143 | |
BC144019.1 | - | 24 - 25 | 2162 | 143 | |
D67035.1 | - | 24 - 25 | 2209 | 143 | |
X95654.1 | - | 24 - 25 | 2136 | 143 |