The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282541.1 | NP_001269470.1 | 28 - 29 | 2628 | 115 |
NM_001282542.1 | NP_001269471.1 | 27 - 28 | 2553 | 115 | |
NM_003176.3 | NP_003167.2 | 28 - 29 | 2625 | 115 | |
NR_104211.1 | 28 - 29 | 2625 | 115 | ||
XM_005271155.3 | XP_005271212.1 | 27 - 28 | 2949 | 115 | |
XM_005271156.1 | XP_005271213.1 | 27 - 28 | 2423 | 115 | |
XM_006710859.1 | XP_006710922.1 | 28 - 29 | 2536 | 115 | |
XM_011542038.2 | XP_011540340.1 | 21 - 22 | 1819 | 115 | |
XM_017002184.1 | XP_016857673.1 | 28 - 29 | 2428 | 115 | |
XM_017002185.1 | XP_016857674.1 | 22 - 23 | 1854 | 115 | |
GenBank mRNA | AK315268.1 | - | 27 - 28 | 2512 | 115 |
BC126266.1 | - | 28 - 29 | 2502 | 115 | |
BC144019.1 | - | 28 - 29 | 2505 | 115 | |
D67035.1 | - | 28 - 29 | 2552 | 115 | |
X95654.1 | - | 28 - 29 | 2479 | 115 |