The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183008.2 | NP_892120.2 | 2 - 3 | 241 | 110 |
GenBank mRNA | AK093258.1 | - | 2 - 3 | 185 | 110 |