The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141969.1 | NP_001135441.1 | 2 - 3 | 408 | 78 |
NM_001141970.1 | NP_001135442.1 | 2 - 3 | 339 | 78 | |
NM_001350.4 | NP_001341.1 | 2 - 3 | 405 | 78 | |
GenBank mRNA | AF015956.2 | - | 1 - 2 | 255 | 78 |
AF039136.1 | - | 2 - 3 | 348 | 78 | |
AF050179.1 | - | 2 - 3 | 316 | 78 | |
AF097742.1 | - | 1 - 2 | 208 | 78 | |
AK292187.1 | - | 2 - 3 | 406 | 78 | |
AK303767.1 | - | 2 - 3 | 282 | 78 | |
AK313034.1 | - | 2 - 3 | 348 | 78 | |
BC109073.1 | - | 2 - 3 | 291 | 78 | |
BC109074.1 | - | 2 - 3 | 291 | 78 | |
CR457085.1 | - | 1 - 2 | 201 | 78 | |
DC345308.1 | - | 2 - 3 | 405 | 78 | |
DC359186.1 | - | 2 - 3 | 408 | 78 | |
HQ436528.1 | - | 1 - 2 | 201 | 78 | |
HQ436529.1 | - | 1 - 2 | 201 | 78 |