Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRCC1
Entrez Gene ID
51315
Gene Name
lysine rich coiled-coil 1
Gene Aliases
CHBP2
Chromosome Location
Chr.2: 88027203 - 88055801 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304526.1 NP_001291455.1 2 - 3 284 142
GenBank mRNA BC015927.1 - 2 - 3 236 142
DB461504.1 - 2 - 3 285 142

View Full Product Details