Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGME1
Entrez Gene ID
92667
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, DDK1, MTDPS11, bA504H3.4
Chromosome Location
Chr.20: 17968594 - 17991122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052865.3 NP_443097.1 2 - 3 819 73
XM_005260870.3 XP_005260927.1 2 - 3 767 73
XM_017028127.1 XP_016883616.1 2 - 3 1041 73
XM_017028128.1 XP_016883617.1 2 - 3 915 73
GenBank mRNA AF306684.1 - 2 - 3 628 73
AF306685.1 - 2 - 3 628 73
AK027503.1 - 2 - 3 623 73
AK315532.1 - 2 - 3 655 73
BC016869.1 - 2 - 3 591 73

View Full Product Details