Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP72
Entrez Gene ID
55722
Gene Name
centrosomal protein 72
Gene Aliases
-
Chromosome Location
Chr.5: 612290 - 663500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018140.3 NP_060610.2 5 - 6 766 73
XM_005248322.3 XP_005248379.1 5 - 6 688 73
XM_011514063.1 XP_011512365.1 5 - 6 712 73
XM_011514064.2 XP_011512366.1 5 - 6 819 73
XM_017009626.1 XP_016865115.1 4 - 5 600 73
XM_017009627.1 XP_016865116.1 4 - 5 560 73
GenBank mRNA AB040952.1 - 5 - 6 704 73
AK001427.1 - 5 - 6 709 73
BC000132.2 - 5 - 6 705 73
BC001750.1 - 5 - 6 692 73

View Full Product Details