Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP72
Entrez Gene ID
55722
Gene Name
centrosomal protein 72
Gene Aliases
-
Chromosome Location
Chr.5: 612290 - 663500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018140.3 NP_060610.2 8 - 9 1410 80
XM_005248322.3 XP_005248379.1 8 - 9 1332 80
XM_011514063.1 XP_011512365.1 8 - 9 1356 80
XM_011514064.2 XP_011512366.1 8 - 9 1463 80
XM_017009626.1 XP_016865115.1 7 - 8 1244 80
XM_017009627.1 XP_016865116.1 7 - 8 1204 80
GenBank mRNA AB040952.1 - 8 - 9 1348 80
AK001427.1 - 8 - 9 1353 80
BC000132.2 - 8 - 9 1349 80
BC001750.1 - 8 - 9 1336 80

View Full Product Details