The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039970.1 | NP_001035059.1 | 11 - 12 | 1553 | 77 |
NM_001278236.1 | NP_001265165.1 | 11 - 12 | 2387 | 77 | |
NM_001278238.1 | NP_001265167.1 | 10 - 11 | 1516 | 77 | |
NM_001278239.1 | NP_001265168.1 | 10 - 11 | 1420 | 77 | |
NM_006906.1 | NP_008837.1 | 11 - 12 | 1649 | 77 | |
NM_032781.3 | NP_116170.3 | 11 - 12 | 1604 | 77 | |
XM_011520411.2 | XP_011518713.1 | 11 - 12 | 1588 | 77 | |
XM_017018434.1 | XP_016873923.1 | 11 - 12 | 1588 | 77 | |
XM_017018435.1 | XP_016873924.1 | 11 - 12 | 1558 | 77 | |
XM_017018436.1 | XP_016873925.1 | 10 - 11 | 1494 | 77 | |
XM_017018437.1 | XP_016873926.1 | 11 - 12 | 1512 | 77 | |
XM_017018438.1 | XP_016873927.1 | 9 - 10 | 1273 | 77 | |
XM_017018439.1 | XP_016873928.1 | 10 - 11 | 1399 | 77 | |
XM_017018440.1 | XP_016873929.1 | 11 - 12 | 1588 | 77 | |
XM_017018441.1 | XP_016873930.1 | 11 - 12 | 1588 | 77 | |
GenBank mRNA | AK090923.1 | - | 11 - 12 | 1513 | 77 |
AK127312.1 | - | 11 - 12 | 2387 | 77 | |
AK294374.1 | - | 10 - 11 | 1256 | 77 | |
AK294673.1 | - | 10 - 11 | 1217 | 77 | |
AK295604.1 | - | 10 - 11 | 1377 | 77 | |
AK316270.1 | - | 10 - 11 | 1301 | 77 | |
AL832541.1 | - | 11 - 12 | 1649 | 77 | |
BC039897.1 | - | 10 - 11 | 1312 | 77 | |
BC046435.1 | - | 1 - 2 | 128 | 77 | |
BC064807.1 | - | 11 - 12 | 1413 | 77 |