The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054022.3 | NP_473363.1 | 6 - 7 | 658 | 59 |
GenBank mRNA | AF229796.1 | - | 6 - 7 | 613 | 59 |