Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
Chromosome Location
Chr.8: 41653225 - 41896762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000037.3 NP_000028.3 5 - 6 511 92
NM_001142446.1 NP_001135918.1 5 - 6 808 92
NM_020475.2 NP_065208.2 5 - 6 511 92
NM_020476.2 NP_065209.2 5 - 6 511 92
NM_020477.2 NP_065210.2 5 - 6 511 92
XM_005273476.4 XP_005273533.1 5 - 6 785 92
XM_011544490.2 XP_011542792.1 6 - 7 707 92
XM_011544491.2 XP_011542793.1 6 - 7 706 92
XM_011544494.2 XP_011542796.1 6 - 7 706 92
XM_011544495.2 XP_011542797.1 6 - 7 705 92
XM_011544496.2 XP_011542798.1 6 - 7 707 92
XM_011544500.2 XP_011542802.1 5 - 6 613 92
XM_011544501.2 XP_011542803.1 5 - 6 613 92
XM_011544502.2 XP_011542804.1 5 - 6 613 92
XM_011544503.2 XP_011542805.1 6 - 7 704 92
XM_011544504.2 XP_011542806.1 5 - 6 613 92
XM_011544505.2 XP_011542807.1 5 - 6 613 92
XM_017013319.1 XP_016868808.1 6 - 7 706 92
XM_017013320.1 XP_016868809.1 6 - 7 708 92
XM_017013321.1 XP_016868810.1 6 - 7 651 92
XM_017013322.1 XP_016868811.1 5 - 6 613 92
XM_017013323.1 XP_016868812.1 6 - 7 627 92
XM_017013324.1 XP_016868813.1 5 - 6 785 92
XM_017013325.1 XP_016868814.1 5 - 6 552 92
XM_017013326.1 XP_016868815.1 5 - 6 545 92
XM_017013327.1 XP_016868816.1 6 - 7 706 92
XM_017013328.1 XP_016868817.1 6 - 7 706 92
XM_017013329.1 XP_016868818.1 5 - 6 783 92
GenBank mRNA AB209418.1 - 5 - 6 808 92
M28880.1 - 5 - 6 511 92
X16609.1 - 5 - 6 511 92

View Full Product Details