Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
Chromosome Location
Chr.8: 41653225 - 41896762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000037.3 NP_000028.3 39 - 40 5477 64
NM_001142446.1 NP_001135918.1 40 - 41 5798 64
NM_020475.2 NP_065208.2 39 - 40 5477 64
NM_020476.2 NP_065209.2 39 - 40 5477 64
NM_020477.2 NP_065210.2 39 - 40 4991 64
XM_005273476.4 XP_005273533.1 40 - 41 5775 64
XM_011544490.2 XP_011542792.1 42 - 43 5742 64
XM_011544491.2 XP_011542793.1 42 - 43 5741 64
XM_011544494.2 XP_011542796.1 41 - 42 5696 64
XM_011544495.2 XP_011542797.1 41 - 42 5695 64
XM_011544496.2 XP_011542798.1 42 - 43 5742 64
XM_011544500.2 XP_011542802.1 39 - 40 5579 64
XM_011544501.2 XP_011542803.1 39 - 40 5579 64
XM_011544502.2 XP_011542804.1 39 - 40 5579 64
XM_011544503.2 XP_011542805.1 41 - 42 5208 64
XM_011544504.2 XP_011542806.1 39 - 40 5093 64
XM_011544505.2 XP_011542807.1 39 - 40 5093 64
XM_017013319.1 XP_016868808.1 41 - 42 5717 64
XM_017013320.1 XP_016868809.1 42 - 43 5743 64
XM_017013321.1 XP_016868810.1 42 - 43 5686 64
XM_017013322.1 XP_016868811.1 41 - 42 5648 64
XM_017013323.1 XP_016868812.1 42 - 43 5662 64
XM_017013324.1 XP_016868813.1 40 - 41 5775 64
XM_017013325.1 XP_016868814.1 41 - 42 5587 64
XM_017013326.1 XP_016868815.1 40 - 41 5535 64
XM_017013327.1 XP_016868816.1 42 - 43 5255 64
XM_017013328.1 XP_016868817.1 41 - 42 5210 64
XM_017013329.1 XP_016868818.1 40 - 41 5287 64
GenBank mRNA AB209418.1 - 40 - 41 5798 64
AK126647.1 - 23 - 24 2871 64
BX647936.1 - 17 - 18 2790 64
M28880.1 - 39 - 40 5477 64
X16609.1 - 39 - 40 5477 64

View Full Product Details