The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284222.1 | NP_001271151.1 | 2 - 3 | 203 | 158 |
NM_018091.5 | NP_060561.3 | 2 - 3 | 268 | 158 | |
XM_006716354.2 | XP_006716417.1 | 2 - 3 | 222 | 158 | |
XM_017013604.1 | XP_016869093.1 | 2 - 3 | 268 | 158 | |
GenBank mRNA | AK001284.1 | - | 2 - 3 | 195 | 158 |
AK022626.1 | - | 2 - 3 | 224 | 158 | |
AL834273.1 | - | 2 - 3 | 126 | 158 | |
BC001240.1 | - | 2 - 3 | 158 | 158 | |
BX648011.1 | - | 2 - 3 | 203 | 158 | |
DA787116.1 | - | 2 - 3 | 268 | 158 | |
DA877844.1 | - | 2 - 3 | 203 | 158 |