Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRPH
Entrez Gene ID
5630
Gene Name
peripherin
Gene Aliases
NEF4, PRPH1
Chromosome Location
Chr.12: 49295013 - 49298698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006262.3 NP_006253.2 5 - 6 1075 86
XM_005269025.1 XP_005269082.1 5 - 6 1188 86
GenBank mRNA AK125587.1 - 5 - 6 2499 86
BC032703.1 - 5 - 6 1035 86

View Full Product Details