The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171818.1 | NP_001165289.1 | 6 - 7 | 822 | 69 |
NM_001171819.1 | NP_001165290.1 | 4 - 5 | 495 | 69 | |
NM_006238.4 | NP_006229.1 | 5 - 6 | 726 | 69 | |
NM_177435.2 | NP_803184.1 | 5 - 6 | 726 | 69 | |
XM_005249193.1 | XP_005249250.1 | 5 - 6 | 741 | 69 | |
XM_006715120.1 | XP_006715183.1 | 7 - 8 | 879 | 69 | |
XM_006715123.1 | XP_006715186.1 | 4 - 5 | 629 | 69 | |
XM_011514707.1 | XP_011513009.1 | 7 - 8 | 1476 | 69 | |
XM_011514709.1 | XP_011513011.1 | 7 - 8 | 1450 | 69 | |
XM_011514710.1 | XP_011513012.1 | 6 - 7 | 803 | 69 | |
XM_017010971.1 | XP_016866460.1 | 8 - 9 | 1572 | 69 | |
XM_017010972.1 | XP_016866461.1 | 7 - 8 | 1503 | 69 | |
XM_017010973.1 | XP_016866462.1 | 7 - 8 | 938 | 69 | |
XM_017010974.1 | XP_016866463.1 | 6 - 7 | 863 | 69 | |
XM_017010975.1 | XP_016866464.1 | 4 - 5 | 614 | 69 | |
GenBank mRNA | AB099507.1 | - | 6 - 7 | 785 | 69 |
AB307691.1 | - | 3 - 4 | 417 | 69 | |
AK122614.1 | - | 5 - 6 | 719 | 69 | |
AK291661.1 | - | 5 - 6 | 685 | 69 | |
AK296425.1 | - | 4 - 5 | 454 | 69 | |
AK304482.1 | - | 3 - 4 | 355 | 69 | |
AY919140.1 | - | 3 - 4 | 417 | 69 | |
BC002715.2 | - | 5 - 6 | 675 | 69 | |
BC007578.1 | - | 5 - 6 | 675 | 69 | |
CN332566.1 | - | 2 - 3 | 289 | 69 | |
HQ692865.1 | - | 3 - 4 | 417 | 69 | |
L07592.1 | - | 6 - 7 | 754 | 69 |