The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289174.1 | NP_001276103.1 | 12 - 13 | 2086 | 74 |
NM_001289175.1 | NP_001276104.1 | 13 - 14 | 1371 | 74 | |
NM_001289176.1 | NP_001276105.1 | 11 - 12 | 1179 | 74 | |
NM_022553.5 | NP_072047.4 | 13 - 14 | 1690 | 74 | |
XM_011514797.1 | XP_011513099.1 | 12 - 13 | 1569 | 74 | |
XM_011514798.2 | XP_011513100.1 | 12 - 13 | 1459 | 74 | |
XM_011514799.1 | XP_011513101.1 | 12 - 13 | 1275 | 74 | |
XM_017011177.1 | XP_016866666.1 | 13 - 14 | 1625 | 74 | |
XM_017011178.1 | XP_016866667.1 | 13 - 14 | 1372 | 74 | |
GenBank mRNA | AK001725.1 | - | 11 - 12 | 1176 | 74 |
AK093057.1 | - | 12 - 13 | 1997 | 74 | |
AK222867.1 | - | 13 - 14 | 1416 | 74 | |
AK297935.1 | - | 13 - 14 | 1346 | 74 | |
AK299563.1 | - | 14 - 15 | 1526 | 74 | |
AL390171.1 | - | 6 - 7 | 639 | 74 | |
AX775737.1 | - | 13 - 14 | 1534 | 74 | |
BC032108.1 | - | 13 - 14 | 1552 | 74 | |
BC040114.1 | - | 13 - 14 | 1571 | 74 |