Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS52
Entrez Gene ID
6293
Gene Name
VPS52, GARP complex subunit
Gene Aliases
ARE1, SAC2, SACM2L, dJ1033B10.5
Chromosome Location
Chr.6: 33250272 - 33271965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289174.1 NP_001276103.1 15 - 16 2417 88
NM_001289175.1 NP_001276104.1 16 - 17 1702 88
NM_001289176.1 NP_001276105.1 14 - 15 1510 88
NM_022553.5 NP_072047.4 16 - 17 2021 88
XM_011514797.1 XP_011513099.1 15 - 16 1900 88
XM_011514798.2 XP_011513100.1 15 - 16 1790 88
XM_011514799.1 XP_011513101.1 15 - 16 1606 88
XM_017011177.1 XP_016866666.1 16 - 17 1956 88
XM_017011178.1 XP_016866667.1 16 - 17 1703 88
GenBank mRNA AK001725.1 - 14 - 15 1507 88
AK093057.1 - 15 - 16 2328 88
AK222867.1 - 16 - 17 1747 88
AK297935.1 - 16 - 17 1677 88
AK299563.1 - 17 - 18 1857 88
AL390171.1 - 9 - 10 970 88
AX775737.1 - 16 - 17 1865 88
BC032108.1 - 16 - 17 1883 88
BC040114.1 - 16 - 17 1902 88

View Full Product Details