The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289174.1 | NP_001276103.1 | 16 - 17 | 2478 | 88 |
NM_001289175.1 | NP_001276104.1 | 17 - 18 | 1763 | 88 | |
NM_001289176.1 | NP_001276105.1 | 15 - 16 | 1571 | 88 | |
NM_022553.5 | NP_072047.4 | 17 - 18 | 2082 | 88 | |
XM_011514797.1 | XP_011513099.1 | 16 - 17 | 1961 | 88 | |
XM_011514798.2 | XP_011513100.1 | 16 - 17 | 1851 | 88 | |
XM_011514799.1 | XP_011513101.1 | 16 - 17 | 1667 | 88 | |
XM_017011178.1 | XP_016866667.1 | 17 - 18 | 1764 | 88 | |
GenBank mRNA | AJ006026.1 | - | 1 - 2 | 72 | 88 |
AK001725.1 | - | 15 - 16 | 1568 | 88 | |
AK093057.1 | - | 16 - 17 | 2389 | 88 | |
AK222867.1 | - | 17 - 18 | 1808 | 88 | |
AK297935.1 | - | 17 - 18 | 1738 | 88 | |
AK299563.1 | - | 18 - 19 | 1918 | 88 | |
AL390171.1 | - | 10 - 11 | 1031 | 88 | |
AX775737.1 | - | 17 - 18 | 1926 | 88 | |
BC032108.1 | - | 17 - 18 | 1944 | 88 | |
BC040114.1 | - | 17 - 18 | 1963 | 88 |