The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289174.1 | NP_001276103.1 | 5 - 6 | 1237 | 67 |
NM_001289175.1 | NP_001276104.1 | 6 - 7 | 522 | 67 | |
NM_001289176.1 | NP_001276105.1 | 6 - 7 | 522 | 67 | |
NM_022553.5 | NP_072047.4 | 6 - 7 | 841 | 67 | |
XM_011514797.1 | XP_011513099.1 | 5 - 6 | 720 | 67 | |
XM_011514798.2 | XP_011513100.1 | 5 - 6 | 610 | 67 | |
XM_011514799.1 | XP_011513101.1 | 5 - 6 | 426 | 67 | |
XM_017011177.1 | XP_016866666.1 | 6 - 7 | 776 | 67 | |
XM_017011178.1 | XP_016866667.1 | 6 - 7 | 523 | 67 | |
XM_017011179.1 | XP_016866668.1 | 6 - 7 | 776 | 67 | |
GenBank mRNA | AK001725.1 | - | 6 - 7 | 519 | 67 |
AK093057.1 | - | 5 - 6 | 1148 | 67 | |
AK222867.1 | - | 6 - 7 | 567 | 67 | |
AK297935.1 | - | 6 - 7 | 497 | 67 | |
AK299563.1 | - | 7 - 8 | 677 | 67 | |
AK300308.1 | - | 6 - 7 | 782 | 67 | |
AK300430.1 | - | 6 - 7 | 539 | 67 | |
AK304080.1 | - | 6 - 7 | 745 | 67 | |
AL559461.3 | - | 6 - 7 | 527 | 67 | |
AX775737.1 | - | 6 - 7 | 685 | 67 | |
BC032108.1 | - | 6 - 7 | 703 | 67 | |
BC040114.1 | - | 6 - 7 | 722 | 67 |