The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003632.2 | NP_003623.1 | 11 - 12 | 1949 | 62 |
XM_005257748.4 | XP_005257805.1 | 11 - 12 | 2714 | 62 | |
XM_017025238.1 | XP_016880727.1 | 13 - 14 | 3084 | 62 | |
GenBank mRNA | U87223.1 | - | 11 - 12 | 1950 | 62 |