Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTNAP1
Entrez Gene ID
8506
Gene Name
contactin associated protein 1
Gene Aliases
CASPR, CNTNAP, NRXN4, P190
Chromosome Location
Chr.17: 42679963 - 42699993 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003632.2 NP_003623.1 3 - 4 581 96
XM_005257748.4 XP_005257805.1 3 - 4 1346 96
XM_017025238.1 XP_016880727.1 5 - 6 1716 96
GenBank mRNA U87223.1 - 3 - 4 582 96

View Full Product Details