The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016640.3 | NP_057724.2 | 2 - 3 | 791 | 131 |
GenBank mRNA | AF146192.2 | - | 2 - 3 | 791 | 131 |
AF217523.1 | - | 2 - 3 | 485 | 131 | |
AK074777.1 | - | 2 - 3 | 759 | 131 | |
AK222704.1 | - | 2 - 3 | 769 | 131 | |
AK222894.1 | - | 2 - 3 | 761 | 131 | |
AL136706.1 | - | 2 - 3 | 764 | 131 | |
AL355715.1 | - | 2 - 3 | 791 | 131 | |
AL355716.1 | - | 2 - 3 | 484 | 131 | |
BC002460.2 | - | 2 - 3 | 742 | 131 | |
BC007735.2 | - | 2 - 3 | 768 | 131 |