The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016640.3 | NP_057724.2 | 3 - 4 | 891 | 138 |
GenBank mRNA | AF146192.2 | - | 3 - 4 | 891 | 138 |
AF217523.1 | - | 3 - 4 | 585 | 138 | |
AK074777.1 | - | 3 - 4 | 859 | 138 | |
AK222704.1 | - | 3 - 4 | 869 | 138 | |
AK222894.1 | - | 3 - 4 | 861 | 138 | |
AL136706.1 | - | 3 - 4 | 864 | 138 | |
AL355715.1 | - | 3 - 4 | 891 | 138 | |
AL355716.1 | - | 3 - 4 | 584 | 138 | |
BC002460.2 | - | 3 - 4 | 842 | 138 | |
BC007735.2 | - | 3 - 4 | 868 | 138 |