The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016640.3 | NP_057724.2 | 4 - 5 | 1066 | 88 |
GenBank mRNA | AF146192.2 | - | 4 - 5 | 1066 | 88 |
AF217523.1 | - | 4 - 5 | 760 | 88 | |
AK074777.1 | - | 4 - 5 | 1034 | 88 | |
AK222704.1 | - | 4 - 5 | 1044 | 88 | |
AK222894.1 | - | 4 - 5 | 1036 | 88 | |
AL136706.1 | - | 4 - 5 | 1039 | 88 | |
AL355715.1 | - | 4 - 5 | 1066 | 88 | |
AL355716.1 | - | 4 - 5 | 759 | 88 | |
BC002460.2 | - | 4 - 5 | 1017 | 88 | |
BC007735.2 | - | 4 - 5 | 1043 | 88 |