The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004719.2 | NP_004710.2 | 11 - 12 | 3845 | 72 |
XM_005269230.2 | XP_005269287.2 | 11 - 12 | 3735 | 72 | |
XM_011538984.2 | XP_011537286.1 | 12 - 13 | 5364 | 72 | |
XM_011538985.1 | XP_011537287.1 | 11 - 12 | 3679 | 72 | |
XM_017020217.1 | XP_016875706.1 | 12 - 13 | 3723 | 72 | |
XM_017020218.1 | XP_016875707.1 | 12 - 13 | 3723 | 72 | |
XM_017020219.1 | XP_016875708.1 | 12 - 13 | 3922 | 72 | |
XM_017020220.1 | XP_016875709.1 | 12 - 13 | 3723 | 72 | |
XM_017020221.1 | XP_016875710.1 | 10 - 11 | 4052 | 72 | |
XM_017020222.1 | XP_016875711.1 | 9 - 10 | 4022 | 72 | |
XM_017020223.1 | XP_016875712.1 | 12 - 13 | 3945 | 72 | |
GenBank mRNA | AF030234.1 | - | 11 - 12 | 3793 | 72 |
AK314093.1 | - | 5 - 6 | 3108 | 72 | |
Y11251.1 | - | 1 - 2 | 3823 | 72 |