The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004719.2 | NP_004710.2 | 14 - 15 | 4521 | 139 |
XM_005269230.2 | XP_005269287.2 | 14 - 15 | 4411 | 139 | |
XM_011538984.2 | XP_011537286.1 | 15 - 16 | 6040 | 139 | |
XM_011538985.1 | XP_011537287.1 | 14 - 15 | 4355 | 139 | |
XM_017020217.1 | XP_016875706.1 | 15 - 16 | 4399 | 139 | |
XM_017020219.1 | XP_016875708.1 | 15 - 16 | 4598 | 139 | |
XM_017020220.1 | XP_016875709.1 | 14 - 15 | 4180 | 139 | |
XM_017020221.1 | XP_016875710.1 | 13 - 14 | 4728 | 139 | |
XM_017020222.1 | XP_016875711.1 | 12 - 13 | 4698 | 139 | |
XM_017020223.1 | XP_016875712.1 | 15 - 16 | 4621 | 139 | |
GenBank mRNA | AF030234.1 | - | 14 - 15 | 4469 | 139 |
AK314093.1 | - | 8 - 9 | 3784 | 139 | |
Y11251.1 | - | 4 - 5 | 4499 | 139 |