The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247996.1 | NP_001234925.1 | 23 - 24 | 2393 | 89 |
NM_018482.3 | NP_060952.2 | 22 - 23 | 2291 | 89 | |
XM_005250925.1 | XP_005250982.1 | 23 - 24 | 2300 | 89 | |
XM_006716563.3 | XP_006716626.1 | 23 - 24 | 2119 | 89 | |
XM_006716564.1 | XP_006716627.1 | 24 - 25 | 2278 | 89 | |
XM_006716565.3 | XP_006716628.1 | 22 - 23 | 2115 | 89 | |
XM_006716566.1 | XP_006716629.1 | 23 - 24 | 2300 | 89 | |
XM_011517052.2 | XP_011515354.1 | 23 - 24 | 2116 | 89 | |
XM_011517053.1 | XP_011515355.1 | 23 - 24 | 2322 | 89 | |
XM_017013467.1 | XP_016868956.1 | 21 - 22 | 2137 | 89 | |
XM_017013468.1 | XP_016868957.1 | 22 - 23 | 2361 | 89 | |
XM_017013469.1 | XP_016868958.1 | 22 - 23 | 2291 | 89 | |
GenBank mRNA | AB033075.1 | - | 16 - 17 | 1524 | 89 |
BC137135.1 | - | 21 - 22 | 2088 | 89 | |
BC144493.1 | - | 22 - 23 | 2190 | 89 | |
BX537768.1 | - | 15 - 16 | 1544 | 89 |