The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193520.1 | NP_001180449.1 | 12 - 13 | 1375 | 86 |
NM_001193521.1 | NP_001180450.1 | 11 - 12 | 1366 | 86 | |
NM_001301202.1 | NP_001288131.1 | 11 - 12 | 1211 | 86 | |
NM_004658.2 | NP_004649.2 | 12 - 13 | 1390 | 86 | |
XM_005253950.4 | XP_005254007.1 | 12 - 13 | 1390 | 86 | |
XM_006719641.3 | XP_006719704.1 | 12 - 13 | 1229 | 86 | |
XM_006719642.3 | XP_006719705.1 | 11 - 12 | 1362 | 86 | |
XM_011538852.2 | XP_011537154.1 | 11 - 12 | 1362 | 86 | |
XM_011538853.2 | XP_011537155.1 | 11 - 12 | 1362 | 86 | |
XM_017020028.1 | XP_016875517.1 | 12 - 13 | 1390 | 86 | |
XM_017020029.1 | XP_016875518.1 | 11 - 12 | 1362 | 86 | |
XM_017020030.1 | XP_016875519.1 | 12 - 13 | 1390 | 86 | |
XM_017020031.1 | XP_016875520.1 | 12 - 13 | 1390 | 86 | |
GenBank mRNA | AB208935.1 | - | 12 - 13 | 1360 | 86 |
AF086713.1 | - | 12 - 13 | 1367 | 86 | |
AK294350.1 | - | 12 - 13 | 1390 | 86 | |
AK309562.1 | - | 11 - 12 | 1363 | 86 | |
BC014420.1 | - | 11 - 12 | 1257 | 86 | |
BC093724.1 | - | 11 - 12 | 1116 | 86 | |
BC143261.1 | - | 11 - 12 | 1116 | 86 |