Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
Chromosome Location
Chr.5: 96662037 - 96774683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042440.3 NP_001035905.1 15 - 16 1425 87
NM_001190442.1 NP_001177371.1 16 - 17 1297 87
NM_001284212.2 NP_001271141.1 13 - 14 1076 87
NM_001284213.2 NP_001271142.1 12 - 13 983 87
NM_173060.3 NP_775083.1 14 - 15 1133 87
NR_104285.1 2 - 3 289 87
XM_006714696.3 XP_006714759.1 18 - 19 1439 87
XM_006714697.3 XP_006714760.1 17 - 18 1456 87
XM_006714698.3 XP_006714761.1 16 - 17 1417 87
XM_006714699.3 XP_006714762.1 17 - 18 1380 87
XM_006714700.3 XP_006714763.1 16 - 17 1399 87
XM_006714701.3 XP_006714764.1 16 - 17 1415 87
XM_006714702.3 XP_006714765.1 16 - 17 1291 87
XM_006714703.3 XP_006714766.1 15 - 16 1376 87
XM_006714704.3 XP_006714767.1 16 - 17 1324 87
XM_006714705.3 XP_006714768.1 15 - 16 1333 87
XM_006714706.3 XP_006714769.1 15 - 16 1251 87
XM_006714707.3 XP_006714770.1 15 - 16 1202 87
XM_006714708.3 XP_006714771.1 14 - 15 1162 87
XM_006714709.3 XP_006714772.1 14 - 15 1125 87
XM_006714710.3 XP_006714773.1 13 - 14 1080 87
XM_006714711.3 XP_006714774.1 13 - 14 1125 87
XM_006714712.3 XP_006714775.1 12 - 13 1072 87
XM_006714713.3 XP_006714776.1 2 - 3 289 87
XM_006714714.3 XP_006714777.1 2 - 3 289 87
XM_006714715.3 XP_006714778.1 3 - 4 1015 87
XM_011543654.2 XP_011541956.1 16 - 17 1375 87
XM_011543656.2 XP_011541958.1 15 - 16 1319 87
XM_011543657.2 XP_011541959.1 15 - 16 1201 87
XM_011543658.2 XP_011541960.1 13 - 14 1054 87
XM_017009911.1 XP_016865400.1 16 - 17 1417 87
XM_017009912.1 XP_016865401.1 17 - 18 1428 87
XM_017009913.1 XP_016865402.1 17 - 18 1397 87
XM_017009914.1 XP_016865403.1 16 - 17 1399 87
XM_017009915.1 XP_016865404.1 17 - 18 1401 87
XM_017009916.1 XP_016865405.1 16 - 17 1415 87
XM_017009917.1 XP_016865406.1 16 - 17 1333 87
XM_017009918.1 XP_016865407.1 15 - 16 1360 87
XM_017009920.1 XP_016865409.1 15 - 16 1243 87
XM_017009921.1 XP_016865410.1 15 - 16 1260 87
XM_017009922.1 XP_016865411.1 14 - 15 1294 87
XM_017009923.1 XP_016865412.1 14 - 15 1294 87
XM_017009925.1 XP_016865414.1 14 - 15 1212 87
XM_017009926.1 XP_016865415.1 14 - 15 1212 87
XM_017009927.1 XP_016865416.1 15 - 16 1201 87
XM_017009928.1 XP_016865417.1 14 - 15 1200 87
XM_017009929.1 XP_016865418.1 14 - 15 1119 87
XM_017009930.1 XP_016865419.1 13 - 14 1086 87
XM_017009931.1 XP_016865420.1 13 - 14 1080 87
XM_017009932.1 XP_016865421.1 12 - 13 1143 87
GenBank mRNA AB208816.1 - 16 - 17 1261 87
AF327443.1 - 11 - 12 975 87
AK292289.1 - 2 - 3 289 87
AK292356.1 - 2 - 3 262 87
AK296919.1 - 16 - 17 1255 87
AK297879.1 - 14 - 15 1075 87
AK298541.1 - 15 - 16 1202 87
AK299384.1 - 15 - 16 1106 87
AK299402.1 - 15 - 16 1182 87
AK302206.1 - 3 - 4 289 87
AK303836.1 - 8 - 9 656 87
AK303900.1 - 11 - 12 869 87
AK311585.1 - 3 - 4 467 87
AK311598.1 - 3 - 4 343 87
AK314290.1 - 13 - 14 1078 87
AK315146.1 - 13 - 14 1077 87
AK315673.1 - 2 - 3 237 87
AL832174.1 - 14 - 15 1118 87
AL832349.1 - 14 - 15 1121 87
AY189741.1 - 4 - 5 288 87
BC013579.1 - 15 - 16 1128 87
BC078149.1 - 10 - 11 711 87
BT009783.1 - 12 - 13 910 87
BX648397.1 - 13 - 14 1027 87
D16217.1 - 15 - 16 1195 87
DB033043.1 - 2 - 3 237 87
U26724.2 - 6 - 7 558 87
U31345.1 - 4 - 5 305 87
U58996.2 - 2 - 3 268 87

View Full Product Details