Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CAST
- Entrez Gene ID
-
831
- Gene Name
- calpastatin
- Gene Aliases
-
BS-17, PLACK
- Chromosome Location
-
Chr.5: 96662037 - 96774683 on Build GRCh38
View Full Product Details