The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184729.2 | NP_001171658.1 | 3 - 4 | 583 | 68 |
NM_015937.5 | NP_057021.2 | 3 - 4 | 583 | 68 | |
NR_047691.1 | 3 - 4 | 543 | 68 | ||
NR_047694.1 | 2 - 3 | 405 | 68 | ||
GenBank mRNA | AB057724.1 | - | 3 - 4 | 493 | 68 |
AF132940.1 | - | 3 - 4 | 487 | 68 | |
AK075469.1 | - | 3 - 4 | 507 | 68 | |
AK225517.1 | - | 3 - 4 | 513 | 68 | |
AK293686.1 | - | 2 - 3 | 328 | 68 | |
AK311492.1 | - | 3 - 4 | 478 | 68 | |
AK316358.1 | - | 3 - 4 | 469 | 68 | |
AL121742.1 | - | 3 - 4 | 486 | 68 | |
AY358588.1 | - | 3 - 4 | 490 | 68 | |
BC015022.1 | - | 3 - 4 | 512 | 68 | |
BC110892.1 | - | 3 - 4 | 491 | 68 | |
BC136827.1 | - | 3 - 4 | 521 | 68 | |
BC136828.1 | - | 3 - 4 | 521 | 68 | |
DA037291.1 | - | 3 - 4 | 566 | 68 |