The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_023068.3 | NP_075556.1 | 3 - 4 | 702 | 67 | |
XM_006723610.3 | XP_006723673.1 | 4 - 5 | 897 | 67 | ||
XM_011529324.2 | XP_011527626.1 | 4 - 5 | 894 | 67 | ||
XM_011529325.2 | XP_011527627.1 | 4 - 5 | 900 | 67 | ||
XM_011529326.2 | XP_011527628.1 | 4 - 5 | 896 | 67 | ||
XM_011529327.2 | XP_011527629.1 | 3 - 4 | 830 | 67 | ||
XM_011529328.2 | XP_011527630.1 | 4 - 5 | 842 | 67 | ||
XM_011529329.1 | XP_011527631.1 | 3 - 4 | 2083 | 67 | ||
XM_011529330.1 | XP_011527632.1 | 3 - 4 | 2083 | 67 | ||
XM_011529331.1 | XP_011527633.1 | 3 - 4 | 2001 | 67 | ||
XM_011529332.1 | XP_011527634.1 | 2 - 3 | 287 | 67 | ||
XM_011529333.2 | XP_011527635.1 | 3 - 4 | 2083 | 67 | ||
GenBank mRNA | AF230073.1 | - | 3 - 4 | 702 | 67 | |
AY927519.1 | - | LIT3529 AY927519 |
2 - 3 | 610 | 67 | |
BC141884.1 | - | 3 - 4 | 702 | 67 |