Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
Chromosome Location
Chr.20: 3732670 - 3753111 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197327.1 NP_001184256.1 12 - 13 1549 79
NM_001318322.1 NP_001305251.1 11 - 12 1454 79
NM_052970.4 NP_443202.3 12 - 13 1552 79
XM_011529151.2 XP_011527453.1 9 - 10 1250 79
XM_017027632.1 XP_016883121.1 12 - 13 1467 79
GenBank mRNA AK056712.1 - 12 - 13 1489 79
AK297120.1 - 10 - 11 1345 79
AK301294.1 - 11 - 12 1420 79
BC090857.1 - 11 - 12 1413 79
BC110881.1 - 12 - 13 1501 79
BC117284.1 - 12 - 13 1450 79
BC126263.1 - 12 - 13 1450 79
BC143932.1 - 12 - 13 1447 79

View Full Product Details