Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU1
Entrez Gene ID
10367
Gene Name
mitochondrial calcium uptake 1
Gene Aliases
CALC, CBARA1, EFHA3, MPXPS
Chromosome Location
Chr.10: 72367326 - 72626191 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195518.1 NP_001182447.1 2 - 3 345 110
NM_006077.3 NP_006068.2 2 - 3 345 110
XM_005269383.2 XP_005269440.1 2 - 3 249 110
XM_011539119.1 XP_011537421.1 2 - 3 249 110
GenBank mRNA AK023318.1 - 2 - 3 264 110
AK225298.1 - 2 - 3 249 110
AK225695.1 - 2 - 3 233 110
BC004190.2 - 2 - 3 257 110
BC004216.2 - 2 - 3 257 110
BC016641.1 - 2 - 3 220 110
DB030067.1 - 2 - 3 295 110

View Full Product Details