Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACE1
Entrez Gene ID
246269
Gene Name
lactation elevated 1
Gene Aliases
AFG1, c222389
Chromosome Location
Chr.6: 108294874 - 108525784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323005.1 NP_001309934.1 3 - 4 618 152
NM_145315.4 NP_660358.2 3 - 4 618 152
XM_005266885.3 XP_005266942.1 4 - 5 1976 152
XM_011535659.2 XP_011533961.1 3 - 4 501 152
XM_011535660.2 XP_011533962.1 3 - 4 501 152
XM_011535661.2 XP_011533963.1 3 - 4 501 152
XM_017010647.1 XP_016866136.1 4 - 5 468 152
GenBank mRNA AF520418.1 - 3 - 4 598 152
BC018445.1 - 3 - 4 598 152

View Full Product Details