Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM29
Entrez Gene ID
23650
Gene Name
tripartite motif containing 29
Gene Aliases
ATDC
Chromosome Location
Chr.11: 120111286 - 120138179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012101.3 NP_036233.2 4 - 5 1461 72
XM_005271490.1 XP_005271547.1 4 - 5 638 72
XM_011542729.1 XP_011541031.1 4 - 5 1485 72
XM_011542732.1 XP_011541034.1 4 - 5 1485 72
XM_011542733.1 XP_011541035.1 4 - 5 638 72
XM_017017453.1 XP_016872942.1 4 - 5 794 72
XM_017017454.1 XP_016872943.1 4 - 5 794 72
GenBank mRNA AF230388.1 - 4 - 5 1461 72
AF230389.1 - 4 - 5 1461 72
AK122720.1 - 5 - 6 1277 72
AK223090.1 - 4 - 5 1419 72
AK299459.1 - 4 - 5 739 72
AK299472.1 - 4 - 5 818 72
AK303969.1 - 4 - 5 699 72
BC010644.1 - 4 - 5 1407 72
BC017352.2 - 4 - 5 1400 72
L24203.1 - 4 - 5 1461 72

View Full Product Details