Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT46
Entrez Gene ID
56912
Gene Name
intraflagellar transport 46
Gene Aliases
C11orf2, C11orf60, CFAP32
Chromosome Location
Chr.11: 118544528 - 118575624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168618.1 NP_001162089.1 7 - 8 902 99
NM_020153.3 NP_064538.3 8 - 9 1055 99
XM_011542905.2 XP_011541207.1 8 - 9 3797 99
XM_011542906.2 XP_011541208.1 7 - 8 3639 99
XM_017018017.1 XP_016873506.1 7 - 8 794 99
XM_017018018.1 XP_016873507.1 6 - 7 765 99
GenBank mRNA AJ249980.1 - 7 - 8 829 99
AK025480.1 - 8 - 9 1014 99
AK289521.1 - 7 - 8 753 99
AL136934.1 - 7 - 8 737 99
BC011647.2 - 7 - 8 740 99
BC012802.2 - 7 - 8 833 99
BC022856.1 - 7 - 8 863 99
CR457344.1 - 5 - 6 483 99

View Full Product Details