The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105515.2 | NP_001098985.1 | 9 - 10 | 1393 | 88 |
NM_001301829.1 | NP_001288758.1 | 9 - 10 | 1393 | 88 | |
NM_001301830.1 | NP_001288759.1 | 8 - 9 | 1168 | 88 | |
NM_005845.4 | NP_005836.2 | 9 - 10 | 1393 | 88 | |
XM_005254025.2 | XP_005254082.1 | 10 - 11 | 1453 | 88 | |
XM_017020319.1 | XP_016875808.1 | 10 - 11 | 1539 | 88 | |
XM_017020320.1 | XP_016875809.1 | 9 - 10 | 1497 | 88 | |
XM_017020322.1 | XP_016875811.1 | 10 - 11 | 1402 | 88 | |
GenBank mRNA | AB208973.1 | - | 7 - 8 | 1012 | 88 |
AF071202.1 | - | 9 - 10 | 1376 | 88 | |
AF541977.1 | - | 9 - 10 | 1266 | 88 | |
AK290317.1 | - | 9 - 10 | 1393 | 88 | |
AK296247.1 | - | 8 - 9 | 1155 | 88 | |
AK298953.1 | - | 9 - 10 | 1357 | 88 | |
AY081219.1 | - | 9 - 10 | 1282 | 88 | |
AY133678.1 | - | 10 - 11 | 1435 | 88 | |
AY133679.1 | - | 11 - 12 | 1548 | 88 | |
AY133680.1 | - | 10 - 11 | 1395 | 88 | |
AY207008.1 | - | 9 - 10 | 1266 | 88 | |
BC041560.1 | - | 9 - 10 | 1363 | 88 |