Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
6737
Gene Name
tripartite motif containing 21
Gene Aliases
RNF81, RO52, Ro/SSA, SSA, SSA1
Chromosome Location
Chr.11: 4384897 - 4393696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003141.3 NP_003132.2 4 - 5 842 112
GenBank mRNA AY742713.1 - 3 - 4 729 112
BC010861.1 - 4 - 5 813 112
M34551.1 - 3 - 4 767 112
M62800.1 - 4 - 5 814 112

View Full Product Details