The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017550.1 | NP_060020.1 | 6 - 7 | 597 | 93 |
XM_005259582.4 | XP_005259639.1 | 6 - 7 | 1181 | 93 | |
XM_006722769.3 | XP_006722832.1 | 6 - 7 | 1185 | 93 | |
XM_011528072.2 | XP_011526374.1 | 6 - 7 | 1183 | 93 | |
XM_011528073.1 | XP_011526375.1 | 6 - 7 | 599 | 93 | |
XM_011528074.2 | XP_011526376.1 | 6 - 7 | 1187 | 93 | |
XM_011528075.2 | XP_011526377.1 | 6 - 7 | 1704 | 93 | |
XM_011528077.2 | XP_011526379.1 | 7 - 8 | 3545 | 93 | |
XM_011528081.2 | XP_011526383.1 | 4 - 5 | 441 | 93 | |
XM_011528082.2 | XP_011526384.1 | 4 - 5 | 372 | 93 | |
GenBank mRNA | AB033019.1 | - | - | 585 | 93 |
BC028203.1 | - | 6 - 7 | 597 | 93 |